Assay Details
Target Gene Details
Entrez Gene ID: | 10656 |
Gene Name: | KH RNA binding domain containing, signal transduction associated 3 |
Gene Aliases: |
Etle, SALP, SLM-2, SLM2, T-STAR, TSTAR, etoile |
Location: |
Chr.8:135457461-135656516 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHDRBS3 | NM_006558.2 | NP_006549.1 | ||
| XM_005250757.2 | XP_005250814.1 | |||
| XM_005250758.3 | XP_005250815.1 | |||
| XM_011516798.1 | XP_011515100.1 | |||
| XM_011516799.1 | XP_011515101.1 | |||
| AF051321.1 | AAC99294.1 | |||
| AF051322.1 | AAC99295.1 | |||
| AF069681.1 | AAC24857.1 | |||
| BC032606.2 | AAH32606.1 | |||
| BC068536.1 | AAH68536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612336 | Chr.8:135449991 - 135459385 on Build GRCh38 | Loss |
|
| nsv612338 | Chr.8:135457139 - 135459385 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79982468] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map