Assay Details
Target Gene Details
Entrez Gene ID: | 28991 |
Gene Name: | COMM domain containing 5 |
Gene Aliases: |
HCARG, HT002 |
Location: |
Chr.8:144837978-144853578 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD5 | NM_001081003.2 | NP_001074472.1 | ||
| NM_001081004.2 | NP_001074473.1 | |||
| NM_001287237.1 | NP_001274166.1 | |||
| NM_014066.3 | NP_054785.2 | |||
| AF113540.1 | AAF14877.1 | |||
| AF290195.1 | AAG09915.1 | |||
| AK001477.1 | BAA91714.1 | |||
| AK023070.1 | BAB14389.1 | |||
| BC002672.2 | AAH02672.1 | |||
| BC003055.1 | AAH03055.1 | |||
| BC065729.1 | AAH65729.1 | |||
| BG705627.1 | ||||
| BI753202.1 | ||||
| BM013651.1 | ||||
| BM924244.1 | ||||
| BU508224.1 | ||||
| H92104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023752 | Chr.8:144835600 - 144866312 on Build GRCh38 | Gain |
|
| nsv469809 | Chr.8:144842323 - 145040108 on Build GRCh38 | Loss |
|
| nsv1031737 | Chr.8:144830982 - 144986209 on Build GRCh38 | Gain |
|
| nsv612969 | Chr.8:144785726 - 144877525 on Build GRCh38 | Loss |
|
| nsv6450 | Chr.8:144850126 - 144882473 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map