Assay Details
Target Gene Details
Entrez Gene ID: | 55626 |
Gene Name: | autophagy and beclin 1 regulator 1 |
Gene Aliases: |
DCAF3, WDR94 |
Location: |
Chr.11:46396412-46594069 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMBRA1 | NM_001267782.1 | NP_001254711.1 | ||
| NM_001267783.1 | NP_001254712.1 | |||
| NM_001300731.1 | NP_001287660.1 | |||
| NM_017749.3 | NP_060219.2 | |||
| XM_005253009.3 | XP_005253066.1 | |||
| XM_005253011.3 | XP_005253068.1 | |||
| XM_005253014.3 | XP_005253071.1 | |||
| XM_006718259.2 | XP_006718322.1 | |||
| XM_006718260.2 | XP_006718323.1 | |||
| XM_017018007.1 | XP_016873496.1 | |||
| AB051523.1 | ||||
| AK000301.1 | BAA91067.1 | |||
| AK023197.1 | BAB14457.1 | |||
| AK057896.1 | ||||
| AK302808.1 | ||||
| AK304530.1 | ||||
| AL834190.1 | ||||
| BC045609.1 | AAH45609.1 | |||
| DQ870924.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626186 | Chr.11:46392885 - 46412875 on Build GRCh38 | Loss |
|
| esv3626185 | Chr.11:46361673 - 46411056 on Build GRCh38 | Loss |
|
| nsv309 | Chr.11:46378128 - 46418953 on Build GRCh38 | Deletion |
|
| esv3626183 | Chr.11:46227941 - 46507334 on Build GRCh38 | Gain |
|
| nsv554202 | Chr.11:46312688 - 46448350 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map