Assay Details
Target Gene Details
Entrez Gene ID: | 23705 |
Gene Name: | cell adhesion molecule 1 |
Gene Aliases: |
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1 |
Location: |
Chr.11:115173625-115504523 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
More Information
Additional Information:
For this assay, SNP(s) [rs151067068] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map