Assay Details
Target Gene Details
Entrez Gene ID: | 6327 |
Gene Name: | sodium voltage-gated channel beta subunit 2 |
Gene Aliases: |
ATFB14 |
Location: |
Chr.11:118162804-118176622 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN2B | NM_004588.4 | NP_004579.1 | ||
| AF007783.1 | AAC26013.1 | |||
| AF049498.1 | AAC05274.1 | |||
| AF107028.1 | AAD47196.1 | |||
| AK308834.1 | ||||
| AK313891.1 | ||||
| AY358945.1 | AAQ89304.1 | |||
| BC036793.1 | AAH36793.1 | |||
| BT019487.1 | AAV38294.1 | |||
| DA290917.1 | ||||
| U87555.1 | AAF21472.1 |
More Information
Set Membership: |
|

Genomic Map