Assay Details
Target Gene Details
Entrez Gene ID: | 9702 |
Gene Name: | centrosomal protein 57 |
Gene Aliases: |
MVA2, PIG8, TSP57 |
Location: |
Chr.11:95790461-95832693 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP57 | NM_001243776.1 | NP_001230705.1 | ||
| NM_001243777.1 | NP_001230706.1 | |||
| NM_014679.4 | NP_055494.2 | |||
| XM_006718945.2 | XP_006719008.1 | |||
| XM_006718946.2 | XP_006719009.1 | |||
| XM_017018592.1 | XP_016874081.1 | |||
| XM_017018593.1 | XP_016874082.1 | |||
| XM_017018594.1 | XP_016874083.1 | |||
| AK098647.1 | ||||
| AK291245.1 | ||||
| AK293277.1 | ||||
| AY225092.1 | AAO73938.1 | |||
| AY239292.1 | AAP72184.1 | |||
| BC001233.2 | AAH01233.1 | |||
| BC029385.1 | ||||
| BC039711.1 | AAH39711.1 | |||
| BG699097.1 | ||||
| D42054.1 | ||||
| DB032301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832235 | Chr.11:95753612 - 95935071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map