Assay Details
Target Gene Details
Entrez Gene ID: | 100526771 |
Gene Name: | TMPRSS4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:118015772-118086793 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS4-AS1 | NR_038318.1 | |||
AB231754.1 | ||||
AB231755.1 | ||||
AB231756.1 | ||||
AB231757.1 |
