Assay Details
Target Gene Details
Entrez Gene ID: | 2900 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 4 |
Gene Aliases: |
EAA1, GRIK, GluK4, KA1 |
Location: |
Chr.11:120511746-120988805 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK4 | NM_001282470.2 | NP_001269399.1 | ||
| NM_001282473.2 | NP_001269402.1 | |||
| NM_014619.4 | NP_055434.2 | |||
| XM_011542784.1 | XP_011541086.1 | |||
| XM_017017621.1 | XP_016873110.1 | |||
| XM_017017622.1 | XP_016873111.1 | |||
| AK292726.1 | ||||
| AK314284.1 | ||||
| BC146652.1 | ||||
| BC150173.1 | ||||
| DA246271.1 | ||||
| S67803.1 | AAB29311.1 |
Target Gene Details
Entrez Gene ID: | 105369532 |
Gene Name: | uncharacterized LOC105369532 |
Gene Aliases: |
- |
Location: |
Chr.11:120811345-120819115 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369532 | NR_133008.1 | |||
| BJ995073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832286 | Chr.11:120715134 - 120875137 on Build GRCh38 | Loss |
|
| nsv1053607 | Chr.11:120670086 - 120931297 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201615547] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map