Assay Details
Target Gene Details
Entrez Gene ID: | 23187 |
Gene Name: | pleckstrin homology like domain family B member 1 |
Gene Aliases: |
LL5A |
Location: |
Chr.11:118606296-118658038 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLDB1 | NM_001144758.2 | NP_001138230.1 | ||
| NM_001144759.2 | NP_001138231.1 | |||
| NM_015157.3 | NP_055972.1 | |||
| XM_006718796.3 | XP_006718859.1 | |||
| XM_011542708.2 | XP_011541010.1 | |||
| XM_011542709.2 | XP_011541011.1 | |||
| AB014538.2 | ||||
| AB094090.1 | BAC76044.1 | |||
| AK074070.2 | ||||
| AK125783.1 | BAC86289.1 | |||
| AK295819.1 | ||||
| AL832373.1 | ||||
| BC098586.1 | AAH98586.1 | |||
| BX489377.1 | ||||
| CR749783.1 | CAH18649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25329 | Chr.11:118608072 - 118612037 on Build GRCh38 | Loss |
|
| nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
|
| nsv556465 | Chr.11:118608595 - 118611488 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map