Assay Details
Target Gene Details
Entrez Gene ID: | 4978 |
Gene Name: | opioid binding protein/cell adhesion molecule like |
Gene Aliases: |
IGLON1, OBCAM, OPCM |
Location: |
Chr.11:132403361-133532983 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPCML | XM_006718846.2 | 8 | 15975 | XP_006718909.1 |
| XM_011542856.2 | 9 | 16288 | XP_011541158.1 |
More Information
Set Membership: |
|

Genomic Map