Assay Details
Target Gene Details
Entrez Gene ID: | 8500 |
Gene Name: | PTPRF interacting protein alpha 1 |
Gene Aliases: |
LIP.1, LIP1, LIPRIN |
Location: |
Chr.11:70270687-70384501 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Additional Information:
For this assay, SNP(s) [rs76773768,rs79787971] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map