Assay Details
Target Gene Details
Entrez Gene ID: | 932 |
Gene Name: | membrane spanning 4-domains A3 |
Gene Aliases: |
CD20L, HTM4 |
Location: |
Chr.11:60056628-60071116 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A3 | NM_001031666.1 | NP_001026836.1 | ||
| NM_001031809.1 | NP_001026979.1 | |||
| NM_006138.4 | NP_006129.4 | |||
| XM_011545363.2 | XP_011543665.1 | |||
| AY095480.1 | AAM23312.1 | |||
| BC008487.1 | AAH08487.1 | |||
| BE564041.1 | ||||
| L35848.1 | AAA62319.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555150 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
| nsv336 | Chr.11:60046173 - 60075342 on Build GRCh38 | Insertion |
|
| nsv509411 | Chr.11:60042589 - 60074584 on Build GRCh38 | Insertion |
|
| nsv832173 | Chr.11:60021411 - 60173087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map