Assay Details
Target Gene Details
Entrez Gene ID: | 2113 |
Gene Name: | ETS proto-oncogene 1, transcription factor |
Gene Aliases: |
ETS-1, EWSR2, c-ets-1, p54 |
Location: |
Chr.11:128458761-128587593 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ETS1 | NM_001143820.1 | NP_001137292.1 | ||
| XM_011542649.2 | XP_011540951.1 | |||
| XM_011542651.1 | XP_011540953.1 | |||
| XM_017017314.1 | XP_016872803.1 | |||
| XM_017017315.1 | XP_016872804.1 | |||
| XM_017017317.1 | XP_016872806.1 | |||
| BX640634.1 | CAE45783.1 |
Target Gene Details
Entrez Gene ID: | 101929517 |
Gene Name: | uncharacterized LOC101929517 |
Gene Aliases: |
- |
Location: |
Chr.11:128526142-128530383 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929517 | NR_120581.1 | |||
| KP742344.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs75386603] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map