Assay Details
Target Gene Details
Entrez Gene ID: | 55031 |
Gene Name: | ubiquitin specific peptidase 47 |
Gene Aliases: |
TRFP |
Location: |
Chr.11:11841423-11959325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP47 | NM_001282659.1 | NP_001269588.1 | ||
NM_017944.3 | NP_060414.3 | |||
XM_005252997.1 | XP_005253054.1 | |||
XM_017017953.1 | XP_016873442.1 | |||
AK027362.1 | BAB55063.1 | |||
AK092290.1 | ||||
AK127264.1 | ||||
AK309240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054113 | Chr.11:11828552 - 11953953 on Build GRCh38 | Loss |
![]() |
nsv553538 | Chr.11:11847002 - 11864294 on Build GRCh38 | Loss |
![]() |
nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832067 | Chr.11:11777158 - 11976774 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv832066 | Chr.11:11684853 - 11865904 on Build GRCh38 | Gain |
![]() ![]() |
dgv1656n54 | Chr.11:11854200 - 11865070 on Build GRCh38 | Loss |
![]() |
esv3579369 | Chr.11:11853475 - 11861994 on Build GRCh38 | Loss |
![]() |
nsv553543 | Chr.11:11854251 - 11870714 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
