Assay Details
Target Gene Details
Entrez Gene ID: | 2313 |
Gene Name: | Fli-1 proto-oncogene, ETS transcription factor |
Gene Aliases: |
EWSR2, SIC-1 |
Location: |
Chr.11:128685263-128813267 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLI1 | NM_001271012.1 | NP_001257941.1 | ||
| XM_017017405.1 | XP_016872894.1 | |||
| AK294279.1 |
Target Gene Details
Entrez Gene ID: | 101929538 |
Gene Name: | uncharacterized LOC101929538 |
Gene Aliases: |
- |
Location: |
Chr.11:128681240-128686922 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929538 | NR_135072.1 | |||
| AK093020.1 |
More Information
Set Membership: |
|

Genomic Map