Assay Details
Target Gene Details
Entrez Gene ID: | 417 |
Gene Name: | ADP-ribosyltransferase 1 |
Gene Aliases: |
ART2, ARTC1, CD296, RT6 |
Location: |
Chr.11:3645110-3665595 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART1 | XM_011520114.2 | 6 | 1991 | XP_011518416.1 |
| XM_017017763.1 | 6 | 2012 | XP_016873252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759796 | Chr.11:3215282 - 3780199 on Build GRCh38 | Gain+Loss |
|
| nsv526441 | Chr.11:3604947 - 3735456 on Build GRCh38 | Gain |
|
| nsv428247 | Chr.11:3595654 - 3780199 on Build GRCh38 | Loss |
|
| nsv832051 | Chr.11:3502717 - 3707484 on Build GRCh38 | Gain |
|
| nsv1046356 | Chr.11:3607607 - 3752985 on Build GRCh38 | Gain |
|
| esv3625147 | Chr.11:3607625 - 3732043 on Build GRCh38 | Gain |
|
| esv3625148 | Chr.11:3609614 - 3680474 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72844389] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map