Assay Details
Target Gene Details
Entrez Gene ID: | 101928847 |
Gene Name: | uncharacterized LOC101928847 |
Gene Aliases: |
- |
Location: |
Chr.11:112959279-112963460 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928847 | NR_120563.1 | 3 | 1310 | |
| AL833634.1 | 3 | 1263 |

Genomic Map