Assay Details
Target Gene Details
Entrez Gene ID: | 84959 |
Gene Name: | ubiquitin associated and SH3 domain containing B |
Gene Aliases: |
STS-1, STS1, TULA-2, TULA2, p70 |
Location: |
Chr.11:122655690-122814479 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBASH3B | NM_032873.4 | NP_116262.2 | ||
XM_011543042.1 | XP_011541344.1 | |||
AB075839.1 | ||||
AK075203.1 | BAC11468.1 | |||
AK222843.1 | BAD96563.1 | |||
AK222846.1 | BAD96566.1 | |||
BC007541.1 | AAH07541.2 | |||
DA197468.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
