Assay Details
Target Gene Details
Entrez Gene ID: | 6484 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 4 |
Gene Aliases: |
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ |
Location: |
Chr.11:126355645-126459601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL4 | NM_001254757.1 | NP_001241686.1 | ||
| NM_006278.2 | NP_006269.1 | |||
| XM_017018134.1 | XP_016873623.1 | |||
| AB009393.1 | ||||
| AL520639.3 | ||||
| AL529887.3 | ||||
| BI259855.1 | ||||
| DA210431.1 | ||||
| X74570.1 | CAA52662.1 |
More Information
Set Membership: |
|

Genomic Map