Assay Details
Target Gene Details
Entrez Gene ID: | 27315 |
Gene Name: | post-GPI attachment to proteins 2 |
Gene Aliases: |
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21 |
Location: |
Chr.11:3797724-3826371 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map