Assay Details
Target Gene Details
Entrez Gene ID: | 8538 |
Gene Name: | BARX homeobox 2 |
Gene Aliases: |
- |
Location: |
Chr.11:129375237-129452279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BARX2 | XM_011543044.2 | XP_011541346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036787 | Chr.11:129345003 - 129455467 on Build GRCh38 | Gain |
|
| nsv952761 | Chr.11:129372806 - 129376705 on Build GRCh38 | Deletion |
|
| nsv1046731 | Chr.11:129228206 - 129415243 on Build GRCh38 | Gain |
|
| nsv1143553 | Chr.11:129375605 - 129375905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map