Assay Details
Target Gene Details
Entrez Gene ID: | 22992 |
Gene Name: | lysine demethylase 2A |
Gene Aliases: |
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA |
Location: |
Chr.11:67119210-67258082 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2A | NM_012308.2 | NP_036440.1 | ||
| NR_027473.1 | ||||
| XM_011544860.2 | XP_011543162.1 | |||
| XM_017017391.1 | XP_016872880.1 | |||
| AB023221.2 | ||||
| AK024505.1 | BAB15795.1 | |||
| BC047486.1 | AAH47486.1 | |||
| BC064360.1 | AAH64360.1 | |||
| JQ710741.1 | ||||
| JQ710742.1 | ||||
| JQ710743.1 | ||||
| JQ710744.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115478046] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map