Assay Details
Target Gene Details
Entrez Gene ID: | 219899 |
Gene Name: | tubulin folding cofactor E like |
Gene Aliases: |
El, LRRC35 |
Location: |
Chr.11:121024094-121090776 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCEL | NM_001130047.1 | NP_001123519.1 | ||
| NM_152715.3 | NP_689928.3 | |||
| XM_017017326.1 | XP_016872815.1 | |||
| AK024239.1 | ||||
| AK295673.1 | ||||
| AK313324.1 | ||||
| AY398644.1 | AAR27875.1 | |||
| BC120988.1 | ||||
| BC120989.1 | ||||
| DB481993.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039908 | Chr.11:121034184 - 121066198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map