Assay Details
Target Gene Details
| Entrez Gene ID: | 5920 | 
| Gene Name: | retinoic acid receptor responder 3 | 
| Gene Aliases: | HRASLS4, HRSL4, PLA1/2-3, RIG1, TIG3 | 
| Location: | Chr.11:63536801-63546458 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 3 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| RARRES3 | NM_004585.4 | NP_004576.2 | ||
| AB030815.1 | BAB08109.1 | |||
| AB453252.1 | ||||
| AF060228.1 | AAC84000.1 | |||
| AF092922.1 | AAF02294.1 | |||
| AK293357.1 | 3 | 817 | ||
| AK312110.1 | ||||
| BC009678.1 | AAH09678.1 | |||
| DA973023.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv468588 | Chr.11:63465891 - 63552767 on Build GRCh38 | Loss |  HRASLS2  RARRES3  HRASLS5  LGALS12 | 
| nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |  HRASLS2  RARRES3  HRASLS5  LGALS12 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top