Assay Details
Target Gene Details
Entrez Gene ID: | 6786 |
Gene Name: | stromal interaction molecule 1 |
Gene Aliases: |
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1 |
Location: |
Chr.11:3855703-4093210 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STIM1 | NM_001277961.1 | NP_001264890.1 | ||
| NM_001277962.1 | NP_001264891.1 | |||
| NM_003156.3 | NP_003147.2 | |||
| AK314928.1 | ||||
| BC021300.2 | AAH21300.1 | |||
| HQ317451.1 | ||||
| JX014264.1 | ||||
| U52426.1 | AAC51627.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78981968] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map