Assay Details
Target Gene Details
Entrez Gene ID: | 80168 |
Gene Name: | monoacylglycerol O-acyltransferase 2 |
Gene Aliases: |
DGAT2L5, DGAT2L5., MGAT2, hDC5 |
Location: |
Chr.11:75717819-75732958 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOGAT2 | NM_025098.3 | 6 | 1658 | NP_079374.2 |
| AK000245.1 | 1 | 867 | ||
| AK128620.1 | 5 | 2115 | BAC87534.1 |
More Information
Set Membership: |
|

Genomic Map