Assay Details
Target Gene Details
Entrez Gene ID: | 11145 |
Gene Name: | phospholipase A2 group XVI |
Gene Aliases: |
AdPLA, H-REV107-1, HRASLS3, HREV107, HREV107-1, HREV107-3, HRSL3 |
Location: |
Chr.11:63574462-63616926 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G16 | NM_001128203.1 | NP_001121675.1 | ||
NM_007069.3 | NP_009000.2 | |||
XM_006718426.1 | XP_006718489.1 | |||
XM_011544741.1 | XP_011543043.1 | |||
AB030814.1 | BAB08108.1 | |||
AB439591.1 | ||||
AF317086.1 | AAL26892.1 | |||
AI590735.1 | ||||
AK313075.1 | ||||
AX775893.1 | CAE11646.1 | |||
BC001387.2 | AAH01387.1 | |||
BC103807.2 | AAI03808.1 | |||
BC103808.2 | AAI03809.1 | |||
BG719903.1 | ||||
CK003948.1 | ||||
X92814.1 | CAA63423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3626646 | Chr.11:63549774 - 63585192 on Build GRCh38 | Gain |
![]() ![]() |
nsv352 | Chr.11:63555836 - 63589650 on Build GRCh38 | Insertion |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
