Assay Details
Target Gene Details
Entrez Gene ID: | 55291 |
Gene Name: | protein phosphatase 6 regulatory subunit 3 |
Gene Aliases: |
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3 |
Location: |
Chr.11:68460712-68615334 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74334702] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map