Assay Details
Target Gene Details
Entrez Gene ID: | 10809 |
Gene Name: | StAR related lipid transfer domain containing 10 |
Gene Aliases: |
CGI-52, NY-CO-28, PCTP2, SDCCAG28 |
Location: |
Chr.11:72754729-72793705 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD10 | NM_006645.2 | NP_006636.2 | ||
| AF151810.1 | AAD34047.1 | |||
| AK095668.1 | ||||
| AK315255.1 | ||||
| BC007919.2 | AAH07919.1 | |||
| BC014033.2 | AAH14033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825992 | Chr.11:72765129 - 72813250 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75134592] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map