Assay Details
Target Gene Details
Entrez Gene ID: | 27315 |
Gene Name: | post-GPI attachment to proteins 2 |
Gene Aliases: |
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21 |
Location: |
Chr.11:3797724-3826371 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Additional Information:
For this assay, SNP(s) [rs76256202] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map