Assay Details
Target Gene Details
Entrez Gene ID: | 2313 |
Gene Name: | Fli-1 proto-oncogene, ETS transcription factor |
Gene Aliases: |
EWSR2, SIC-1 |
Location: |
Chr.11:128685263-128813267 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLI1 | NM_001167681.2 | NP_001161153.1 | ||
| NM_001271010.1 | NP_001257939.1 | |||
| NM_001271012.1 | NP_001257941.1 | |||
| NM_002017.4 | NP_002008.2 | |||
| XM_011542701.2 | XP_011541003.1 | |||
| XM_011542702.1 | XP_011541004.1 | |||
| XM_017017405.1 | XP_016872894.1 | |||
| XM_017017406.1 | XP_016872895.1 | |||
| AK294279.1 | ||||
| AK300153.1 | ||||
| AK313370.1 | ||||
| AY029368.1 | AAK50443.1 | |||
| BC001670.1 | AAH01670.1 | |||
| BC010115.1 | AAH10115.1 | |||
| DC381599.1 | ||||
| M93255.1 | AAA58479.1 | |||
| M98833.3 | AAA35812.2 | |||
| S45205.1 | AAB23637.1 | |||
| X67001.1 | CAA47399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952759 | Chr.11:128696006 - 128745405 on Build GRCh38 | Duplication |
|
| esv3628048 | Chr.11:128701861 - 128712027 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113294851] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map