Assay Details
Target Gene Details
Entrez Gene ID: | 22992 |
Gene Name: | lysine demethylase 2A |
Gene Aliases: |
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA |
Location: |
Chr.11:67119210-67258082 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KDM2A | NM_012308.2 | NP_036440.1 | ||
NR_027473.1 | ||||
XM_011544860.2 | XP_011543162.1 | |||
XM_017017391.1 | XP_016872880.1 | |||
AB023221.2 | ||||
AK024505.1 | BAB15795.1 | |||
BC047486.1 | AAH47486.1 | |||
BC064360.1 | AAH64360.1 | |||
JQ710741.1 | ||||
JQ710742.1 | ||||
JQ710743.1 | ||||
JQ710744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1976n54 | Chr.11:67141279 - 67149366 on Build GRCh38 | Loss |
![]() |
esv2661599 | Chr.11:67139759 - 67149939 on Build GRCh38 | Deletion |
![]() |
nsv555252 | Chr.11:67143355 - 67148921 on Build GRCh38 | Loss |
![]() |
esv3626707 | Chr.11:67139805 - 67149822 on Build GRCh38 | Loss |
![]() |
esv23935 | Chr.11:67139958 - 67149624 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
