Assay Details
Target Gene Details
Entrez Gene ID: | 79703 |
Gene Name: | chromosome 11 open reading frame 80 |
Gene Aliases: |
TOP6BL, TOPOVIBL |
Location: |
Chr.11:66744736-66843516 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf80 | NM_001302084.1 | NP_001289013.1 | ||
| NM_024650.3 | NP_078926.3 | |||
| AK026184.1 | BAB15387.1 | |||
| AK225435.1 | ||||
| AK302025.1 | ||||
| BC028240.1 | AAH28240.1 | |||
| BI559574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516087 | Chr.11:66748578 - 66812101 on Build GRCh38 | Gain |
|
| nsv555225 | Chr.11:66700998 - 66866926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map