Assay Details
Target Gene Details
Entrez Gene ID: | 84959 |
Gene Name: | ubiquitin associated and SH3 domain containing B |
Gene Aliases: |
STS-1, STS1, TULA-2, TULA2, p70 |
Location: |
Chr.11:122655690-122814479 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBASH3B | NM_032873.4 | NP_116262.2 | ||
| XM_011543042.1 | XP_011541344.1 | |||
| AB075839.1 | ||||
| AK075203.1 | BAC11468.1 | |||
| AK222843.1 | BAD96563.1 | |||
| AK222846.1 | BAD96566.1 | |||
| BC007541.1 | AAH07541.2 | |||
| DA197468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053418 | Chr.11:122662739 - 122818935 on Build GRCh38 | Gain |
|
| esv3627945 | Chr.11:122683658 - 122694511 on Build GRCh38 | Loss |
|
| nsv983212 | Chr.11:122685001 - 122695304 on Build GRCh38 | Deletion |
|
| nsv1050579 | Chr.11:122684157 - 122728417 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map