Assay Details
Target Gene Details
Entrez Gene ID: | 8525 |
Gene Name: | diacylglycerol kinase zeta |
Gene Aliases: |
DAGK5, DAGK6, DGK-ZETA, hDGKzeta |
Location: |
Chr.11:46332905-46380554 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKZ | NM_201532.2 | NP_963290.1 | ||
| XM_005253181.3 | XP_005253238.1 | |||
| XM_011520423.1 | XP_011518725.1 | |||
| AK124594.1 | BAC85894.1 | |||
| AK126970.1 | BAC86770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626183 | Chr.11:46227941 - 46507334 on Build GRCh38 | Gain |
|
| nsv554202 | Chr.11:46312688 - 46448350 on Build GRCh38 | Loss |
|
| nsv825867 | Chr.11:46281633 - 46404063 on Build GRCh38 | Gain |
|
| nsv554201 | Chr.11:46260284 - 46401136 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map