Assay Details
Target Gene Details
Entrez Gene ID: | 466 |
Gene Name: | activating transcription factor 1 |
Gene Aliases: |
EWS-ATF1, FUS/ATF-1, TREB36 |
Location: |
Chr.12:50763682-50821162 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF1 | NM_005171.4 | NP_005162.1 | ||
| XM_011538386.2 | XP_011536688.1 | |||
| XM_011538387.2 | XP_011536689.1 | |||
| XM_011538388.2 | XP_011536690.1 | |||
| XM_017019331.1 | XP_016874820.1 | |||
| XM_017019332.1 | XP_016874821.1 | |||
| XM_017019333.1 | XP_016874822.1 | |||
| XM_017019334.1 | XP_016874823.1 | |||
| XM_017019335.1 | XP_016874824.1 | |||
| XM_017019336.1 | XP_016874825.1 | |||
| AK299240.1 | ||||
| AK313406.1 | ||||
| BC029619.1 | AAH29619.1 | |||
| BG502726.1 | ||||
| BG545632.1 | ||||
| BI712653.1 |
More Information
Set Membership: |
|

Genomic Map