Assay Details
Target Gene Details
Entrez Gene ID: | 283316 |
Gene Name: | CD163 molecule like 1 |
Gene Aliases: |
CD163B, M160, SCARI2 |
Location: |
Chr.12:7320510-7444185 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD163L1 | NM_001297650.1 | NP_001284579.1 | ||
| NM_174941.5 | NP_777601.2 | |||
| XM_011520616.1 | XP_011518918.1 | |||
| XM_011520617.2 | XP_011518919.1 | |||
| XM_011520618.2 | XP_011518920.1 | |||
| XM_011520619.1 | XP_011518921.1 | |||
| XM_011520620.1 | XP_011518922.1 | |||
| XM_011520621.2 | XP_011518923.1 | |||
| AF264014.1 | AAF91396.1 | |||
| AK058038.1 | ||||
| AK303372.1 | ||||
| BC104962.1 | AAI04963.1 | |||
| BC104964.1 | AAI04965.1 | |||
| GQ397482.1 |
More Information
Set Membership: |
|

Genomic Map