Assay Details
Target Gene Details
Entrez Gene ID: | 8683 |
Gene Name: | serine and arginine rich splicing factor 9 |
Gene Aliases: |
SFRS9, SRp30c |
Location: |
Chr.12:120461668-120469755 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF9 | NM_003769.2 | NP_003760.1 | ||
| AK290644.1 | ||||
| AK294254.1 | ||||
| AK309778.1 | ||||
| BC093971.1 | AAH93971.1 | |||
| BC093973.1 | AAH93973.1 | |||
| BU786332.1 | ||||
| U30825.1 | AAA93069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509482 | Chr.12:120435117 - 120495113 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map