Assay Details
Target Gene Details
Entrez Gene ID: | 1975 |
Gene Name: | eukaryotic translation initiation factor 4B |
Gene Aliases: |
EIF-4B, PRO1843 |
Location: |
Chr.12:53006258-53042209 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4B | NM_001300821.1 | NP_001287750.1 | ||
| NM_001417.5 | NP_001408.2 | |||
| AB076839.1 | BAB82380.1 | |||
| AK222528.1 | BAD96248.1 | |||
| AK293731.1 | ||||
| AK299329.1 | ||||
| AK299510.1 | ||||
| AK299526.1 | ||||
| AK310244.1 | ||||
| BC073139.1 | AAH73139.1 | |||
| BC073154.1 | AAH73154.1 | |||
| BC098437.1 | AAH98437.1 | |||
| CR456958.1 | CAG33239.1 | |||
| CR627377.1 | CAH10474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map