Assay Details
Target Gene Details
Entrez Gene ID: | 84915 |
Gene Name: | family with sequence similarity 222 member A |
Gene Aliases: |
C12orf34 |
Location: |
Chr.12:109714382-109770509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM222A | NM_032829.2 | NP_116218.2 | ||
| DN994281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2856n54 | Chr.12:109702208 - 109717766 on Build GRCh38 | Gain |
|
| nsv560133 | Chr.12:109712747 - 109732223 on Build GRCh38 | Gain |
|
| nsv560148 | Chr.12:109713735 - 109717766 on Build GRCh38 | Loss |
|
| nsv560142 | Chr.12:109713678 - 109717766 on Build GRCh38 | Gain |
|

Genomic Map