Assay Details
Target Gene Details
Entrez Gene ID: | 100874071 |
Gene Name: | USP12 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:27172259-27183503 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP12-AS2 | NR_046548.1 | |||
| AW449459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2583727 | Chr.13:27175450 - 27176913 on Build GRCh38 | Deletion |
|

Genomic Map