Assay Details
Target Gene Details
Entrez Gene ID: | 6564 |
Gene Name: | solute carrier family 15 member 1 |
Gene Aliases: |
HPECT1, HPEPT1, PEPT1 |
Location: |
Chr.13:98683801-98752675 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC15A1 | NM_005073.3 | NP_005064.1 | ||
AF043233.1 | AAC15477.1 | |||
AY634368.1 | AAT48121.1 | |||
BC096327.1 | AAH96327.1 | |||
BC096328.3 | AAH96328.1 | |||
BC096329.3 | AAH96329.1 | |||
EU544224.1 | ||||
U13173.1 | AAB61693.1 | |||
U21936.1 | AAA63797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1038016 | Chr.13:98665177 - 98817733 on Build GRCh38 | Gain |
![]() ![]() |
esv2747861 | Chr.13:98625149 - 98716478 on Build GRCh38 | Deletion |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
