Assay Details
Target Gene Details
Entrez Gene ID: | 10257 |
Gene Name: | ATP binding cassette subfamily C member 4 |
Gene Aliases: |
MOAT-B, MOATB, MRP4 |
Location: |
Chr.13:95019829-95301550 on Build GRCh38 |
Assay Gene Location: | Within Intron 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCC4 | NM_001301829.1 | NP_001288758.1 | ||
NM_005845.4 | NP_005836.2 | |||
XM_005254025.2 | XP_005254082.1 | |||
XM_017020319.1 | XP_016875808.1 | |||
XM_017020321.1 | XP_016875810.1 | |||
AF071202.1 | AAC27076.1 | |||
AF071203.1 | AAC27077.1 | |||
AF541977.1 | AAN17334.1 | |||
AK309460.1 | ||||
AY081219.1 | AAL88745.1 | |||
AY207008.1 | AAO37649.1 | |||
BX647118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525194 | Chr.13:95020696 - 95084938 on Build GRCh38 | Loss |
![]() |
nsv832688 | Chr.13:94980939 - 95127944 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
