Assay Details
Target Gene Details
Entrez Gene ID: | 10443 |
Gene Name: | NEDD4 binding protein 2 like 2 |
Gene Aliases: |
92M18.3, CG005, CG016, PFAAP5 |
Location: |
Chr.13:32432488-32542710 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Target Gene Details
Entrez Gene ID: | 116828 |
Gene Name: | N4BPL2 intronic transcript 2 |
Gene Aliases: |
CG030 |
Location: |
Chr.13:32504506-32509395 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP2L2-IT2 | NR_026928.1 | 1 | 496 | |
| U50531.1 | 1 | 496 |
More Information
Set Membership: |
|

Genomic Map