Assay Details
Target Gene Details
Entrez Gene ID: | 100861550 |
Gene Name: | PDX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:27829759-27921404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDX1-AS1 | NR_047484.1 |

Genomic Map