Assay Details
Target Gene Details
Entrez Gene ID: | 100289274 |
Gene Name: | DNAJC3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:95672835-95676925 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC3-AS1 | NR_132117.1 | 1 | 638 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951908 | Chr.13:95674447 - 95682946 on Build GRCh38 | Duplication |
|

Genomic Map