Assay Details
Target Gene Details
Entrez Gene ID: | 26960 |
Gene Name: | neurobeachin |
Gene Aliases: |
BCL8B, LYST2 |
Location: |
Chr.13:34942287-35672737 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBEA | NM_015678.4 | NP_056493.3 | ||
| XM_005266346.4 | XP_005266403.1 | |||
| XM_005266347.4 | XP_005266404.2 | |||
| XM_005266348.4 | XP_005266405.2 | |||
| XM_006719803.3 | XP_006719866.1 | |||
| XM_006719805.3 | XP_006719868.1 | |||
| XM_006719806.3 | XP_006719869.1 | |||
| XM_011535047.2 | XP_011533349.1 | |||
| XM_017020544.1 | XP_016876033.1 | |||
| XM_017020545.1 | XP_016876034.1 | |||
| XM_017020546.1 | XP_016876035.1 | |||
| AF467288.1 | AAM53531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580599 | Chr.13:34958412 - 34992369 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73490346] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map