Assay Details
Target Gene Details
Entrez Gene ID: | 5925 |
Gene Name: | RB transcriptional corepressor 1 |
Gene Aliases: |
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110 |
Location: |
Chr.13:48303747-48481890 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RB1 | NM_000321.2 | NP_000312.2 | ||
XM_011535171.2 | XP_011533473.1 | |||
AB208788.1 | BAD92025.1 | |||
AK291258.1 | ||||
AK299179.1 | ||||
AK300284.1 | ||||
AK307730.1 | ||||
BC039060.1 | AAH39060.1 | |||
BC040540.1 | AAH40540.1 | |||
L41870.1 | AAB59465.1 | |||
M15400.1 | AAA69807.1 | |||
M19701.1 | AAA60259.1 | |||
M28419.1 | AAA69808.1 | |||
M33647.1 | AAA69806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1648n100 | Chr.13:48324814 - 48351221 on Build GRCh38 | Loss |
![]() |
nsv1042504 | Chr.13:48337128 - 48351221 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
