Assay Details
Target Gene Details
Entrez Gene ID: | 219333 |
Gene Name: | ubiquitin specific peptidase 12 |
Gene Aliases: |
UBH1, USP12L1 |
Location: |
Chr.13:27066150-27171896 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP12 | NM_182488.3 | NP_872294.2 | ||
XM_005266282.2 | XP_005266339.1 | |||
XM_011534956.2 | XP_011533258.1 | |||
XM_017020416.1 | XP_016875905.1 | |||
AF022789.1 | AAC23551.1 | |||
AK289685.1 | ||||
BC026072.1 | AAH26072.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
