Assay Details
Target Gene Details
Entrez Gene ID: | 171425 |
Gene Name: | citrate lyase beta like |
Gene Aliases: |
CLB |
Location: |
Chr.13:99606664-99909459 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLYBL | NM_206808.3 | NP_996531.1 | ||
NR_104592.1 | ||||
XM_005254030.2 | XP_005254087.1 | |||
XM_005254031.2 | XP_005254088.1 | |||
XM_006719915.3 | XP_006719978.1 | |||
XM_011521051.2 | XP_011519353.1 | |||
XM_011521052.2 | XP_011519354.1 | |||
XM_017020402.1 | XP_016875891.1 | |||
XM_017020403.1 | XP_016875892.1 | |||
XM_017020404.1 | XP_016875893.1 | |||
AF428253.1 | AAL84703.1 | |||
AK095506.1 | BAC04561.1 | |||
AK300510.1 | ||||
BC034360.1 | AAH34360.1 | |||
DA621336.1 |
Target Gene Details
Entrez Gene ID: | 101927437 |
Gene Name: | uncharacterized LOC101927437 |
Gene Aliases: |
- |
Location: |
Chr.13:99740870-99957165 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927437 | NR_120421.1 | |||
DA140610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2747875 | Chr.13:99669768 - 100007232 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832696 | Chr.13:99667339 - 99829982 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
